(Iodomethyl)cyclobutane

Supplier: Apollo Scientific

OR304369-1G OR304369-25G OR304369-5G
APOSOR304369-1GEA 273 EUR
APOSOR304369-1G APOSOR304369-25G APOSOR304369-5G
(Iodomethyl)cyclobutane
(Iodomethyl)cyclobutane
Formula: C₅H₉I
MW: 196,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410737
CAS Number: 16408-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR