N-(3-Aminocyclobutyl)acetamide hydrochloride

Supplier: Apollo Scientific

OR304378-1G OR304378-500MG
APOSOR304378-1GEA 1040 EUR
APOSOR304378-1G APOSOR304378-500MG
N-(3-Aminocyclobutyl)acetamide hydrochloride
N-(3-Aminocyclobutyl)acetamide hydrochloride
Formula: C₆H₁₂N₂O·ClH
MW: 164,633 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29059329
CAS Number: 1860028-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR