2,5-Diazaspiro[3.4]octan-6-one hydrochloride

Supplier: Apollo Scientific

OR304396-1G OR304396-250MG OR304396-500MG
APOSOR304396-1GEA 1850 EUR
APOSOR304396-1G APOSOR304396-250MG APOSOR304396-500MG
2,5-Diazaspiro[3.4]octan-6-one hydrochloride
2,5-Diazaspiro[3.4]octan-6-one hydrochloride
Formula: C₆H₁₀N₂O·ClH
MW: 162,617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28155140
CAS Number: 1630906-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR