6-Methoxy-1-benzothiophene-2-carboxylic acid
Supplier: Apollo Scientific
Synonyms:
6-Methoxybenzo[b]thiophene-2-carboxylic acid
OR305047-250MG
OR305047-1G
APOSOR305047-250MGEA
72.5
EUR
APOSOR305047-250MG
APOSOR305047-1G
6-Methoxy-1-benzothiophene-2-carboxylic acid
6-Methoxy-1-benzothiophene-2-carboxylic acid
Formula:
C₁₀H₈O₃S MW: 208,234 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07371542 CAS Number: 102539-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...