5-Bromo-1H-indazole-3-carbaldehyde

Supplier: Apollo Scientific

OR305082-1G OR305082-250MG
APOSOR305082-1GEA 90.2 EUR
APOSOR305082-1G APOSOR305082-250MG
5-Bromo-1H-indazole-3-carbaldehyde
5-Bromo-1H-indazole-3-carbaldehyde
Formula: C₈H₅BrN₂O
MW: 225,042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738287
CAS Number: 201227-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR