4-Chloro-3-methyl-1H-indazole

Supplier: Apollo Scientific
Warning

OR305098-100MG OR305098-250MG OR305098-50MG
APOSOR305098-100MGEA 72.5 EUR
APOSOR305098-100MG APOSOR305098-250MG APOSOR305098-50MG
4-Chloro-3-methyl-1H-indazole
4-Chloro-3-methyl-1H-indazole
Formula: C₈H₇ClN₂
Melting Pt: 176…177 °C
Storage Temperature: Ambient
MDL Number: MFCD13176960
CAS Number: 16640-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR