4-(1H-Imidazol-2-yl)phenol

Supplier: Apollo Scientific

OR305176-1G OR305176-250MG
APOSOR305176-1GEA 459 EUR
APOSOR305176-1G APOSOR305176-250MG
4-(1H-Imidazol-2-yl)phenol
4-(1H-Imidazol-2-yl)phenol
Formula: C₉H₈N₂O
MW: 160,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08668843
CAS Number: 15548-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR