2,3-Dihydro-1H-indol-4-amine

Supplier: Apollo Scientific

OR305260-250MG OR305260-1G OR305260-5G
APOSOR305260-250MGEA 59.6 EUR
APOSOR305260-250MG APOSOR305260-1G APOSOR305260-5G
2,3-Dihydro-1H-indol-4-amine
2,3-Dihydro-1H-indol-4-amine
Formula: C₈H₁₀N₂
Melting Pt: 44…45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08234591
CAS Number: 52537-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR