5-Chloroisoindolin-1-one

Supplier: Apollo Scientific
Warning

OR305284-1G OR305284-250MG
APOSOR305284-1GEA 168 EUR
APOSOR305284-1G APOSOR305284-250MG
5-Chloroisoindolin-1-one
5-Chloroisoindolin-1-one
Formula: C₈H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD08234731
CAS Number: 74572-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR