8-Chloroimidazo[1,5-a]pyrazine 95%
Supplier: Apollo Scientific
|
Warning
|
OR305494-250MG
OR305494-100MG
OR305494-1G
APOSOR305494-250MGEA
173
EUR
APOSOR305494-250MG
APOSOR305494-100MG
APOSOR305494-1G
8-Chloroimidazo[1,5-a]pyrazine 95%
8-Chloroimidazo[1,5-a]pyrazine
Formula:
C₆H₄ClN₃ MW: 153,569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20040069 CAS Number: 56468-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...