5-Methyl-1H-indazol-4-amine 95+%

Supplier: Apollo Scientific
Warning

OR305526-100MG OR305526-1G OR305526-250MG
APOSOR305526-100MGEA 93.1 EUR
APOSOR305526-100MG APOSOR305526-1G APOSOR305526-250MG
5-Methyl-1H-indazol-4-amine 95+%
5-Methyl-1H-indazol-4-amine
Formula: C₈H₉N₃
MW: 147,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19204122
CAS Number: 81115-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR