Isoquinoline-6-carboxaldehyde 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Formylisoquinoline, 6-Formyl-2-azanaphthalene, Isoquinoline-6-carbaldehyde

OR30560-1G OR30560-250MG
APOSOR30560-1GEA 634 EUR
APOSOR30560-1G APOSOR30560-250MG
Isoquinoline-6-carboxaldehyde 95+%
Isoquinoline-6-carboxaldehyde
Formula: C₁₀H₇NO
MW: 157,169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06227438
CAS Number: 173089-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR