1H-Indol-3-ylboronic acid 92%
Supplier: Apollo Scientific
OR305637-250MG
OR305637-1G
APOSOR305637-250MGEA
132
EUR
APOSOR305637-250MG
APOSOR305637-1G
1H-Indol-3-ylboronic acid 92%
1H-Indol-3-ylboronic acid
Formula:
C₈H₈BNO₂ MW: 160,966 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094806 CAS Number: 741253-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...