3-(2-Chlorophenyl)-3-methylbutanoic acid

Supplier: Apollo Scientific
Warning

OR306025-1G OR306025-250MG
APOSOR306025-1GEA 906 EUR
APOSOR306025-1G APOSOR306025-250MG
3-(2-Chlorophenyl)-3-methylbutanoic acid
3-(2-Chlorophenyl)-3-methylbutanoic acid
Formula: C₁₁H₁₃ClO₂
MW: 212.673 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16308461
CAS Number: 91427-34-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR