(Cyclopropylmethyl)hydrazine dihydrochloride

Supplier: Apollo Scientific

OR306202-1G OR306202-250MG
APOSOR306202-1GEA 217 EUR
APOSOR306202-1G APOSOR306202-250MG
(Cyclopropylmethyl)hydrazine dihydrochloride
(Cyclopropylmethyl)hydrazine dihydrochloride
Formula: C₄H₁₀N₂·2ClH
MW: 159,057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09834106
CAS Number: 809282-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR