5-Amino-1,4-dimethyl-1H-pyrazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,4-Dimethyl-1H-pyrazol-5-amine
OR306261-1G
OR306261-5G
APOSOR306261-1GEA
277
EUR
APOSOR306261-1G
APOSOR306261-5G
5-Amino-1,4-dimethyl-1H-pyrazole
5-Amino-1,4-dimethyl-1H-pyrazole
Formula:
C₅H₉N₃ MW: 111,145 g/mol Boiling Pt: 238 °C Melting Pt: 168…170 °C Density: 1,17 g/cm³ (25 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD08059942 CAS Number: 3524-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...