5-Bromo-2,4-dimethylaniline

Supplier: Apollo Scientific
Warning

OR30656-1G OR30656-25G OR30656-5G
APOSOR30656-1GEA 14.7 EUR
APOSOR30656-1G APOSOR30656-25G APOSOR30656-5G
5-Bromo-2,4-dimethylaniline
5-Bromo-2,4-dimethylaniline
Formula: C₈H₁₀BrN
Melting Pt: 96…100 °C
Storage Temperature: Ambient
MDL Number: MFCD09038481
CAS Number: 69383-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR