5-Cyclopropylpicolinic acid

Supplier: Apollo Scientific

OR306657-1G OR306657-250MG
APOSOR306657-1GEA 574 EUR
APOSOR306657-1G APOSOR306657-250MG
5-Cyclopropylpicolinic acid
5-Cyclopropylpicolinic acid
Formula: C₉H₉NO₂
MW: 163,173 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17259279
CAS Number: 1174322-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR