3-Cyclopropylaniline 95%
Supplier: Apollo Scientific
|
Warning
|
OR307256-1G
OR307256-5G
APOSOR307256-1GEA
161
EUR
APOSOR307256-1G
APOSOR307256-5G
3-Cyclopropylaniline 95%
3-Cyclopropylaniline
Formula:
C₉H₁₁N MW: 133,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06802404 CAS Number: 485402-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...