3-tert-Butoxycarbonylamino-3-cyclohexyl-propionic acid
Supplier: Apollo Scientific
|
Warning
|
OR307571-1G
OR307571-250MG
APOSOR307571-1GEA
197
EUR
APOSOR307571-1G
APOSOR307571-250MG
3-tert-Butoxycarbonylamino-3-cyclohexyl-propionic acid
3-tert-Butoxycarbonylamino-3-cyclohexyl-propionic acid
Formula:
C₁₄H₂₅NO₄ MW: 271.353 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03003684 CAS Number: 458529-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...