2-(1,2,3,4-Tetrahydroisoquinolin-1-yl)-1-naphthol

Supplier: Apollo Scientific

OR307665-1G OR307665-250MG
APOSOR307665-1GEA 591 EUR
APOSOR307665-1G APOSOR307665-250MG
2-(1,2,3,4-Tetrahydroisoquinolin-1-yl)-1-naphthol
2-(1,2,3,4-Tetrahydroisoquinolin-1-yl)-1-naphthol
Formula: C₁₉H₁₇NO
MW: 275,344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09881436
CAS Number: 897035-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR