(E)-2-Cyclopropylethylene-1-boronic acid, pinacol ester 96%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(E)-2-Cyclopropylethene-1-boronic acid, (E)-2-(2-Cyclopropylvinyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-Cyclopropylvinylboronic acid, pinacol ester
OR30768-1G
OR30768-25G
OR30768-5G
APOSOR30768-1GEA
101
EUR
APOSOR30768-1G
APOSOR30768-25G
APOSOR30768-5G
(E)-2-Cyclopropylethylene-1-boronic acid, pinacol ester 96%
(E)-2-Cyclopropylethylene-1-boronic acid, pinacol ester
Formula:
C₁₁H₁₉BO₂ MW: 194,078 g/mol Boiling Pt: 90…95 °C (3 mm Hg) Density: 0,937 g/cm³ (25 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD07369776 CAS Number: 849061-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...