Benzofuro[2,3-d]pyridazin-4(3H)-one
Supplier: Apollo Scientific
OR307759-1G
OR307759-250MG
APOSOR307759-1GEA
574
EUR
APOSOR307759-1G
APOSOR307759-250MG
Benzofuro[2,3-d]pyridazin-4(3H)-one
Benzofuro[2,3-d]pyridazin-4(3H)-one
Formula:
C₁₀H₆N₂O₂ MW: 186,167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09997949 CAS Number: 50266-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...