4-Hydroxy-6-methylquinoline

Supplier: Apollo Scientific

Synonyms: 6-Methylquinolin-4-ol

OR307813-1G OR307813-5G
APOSOR307813-1GEA 121 EUR
APOSOR307813-1G APOSOR307813-5G
4-Hydroxy-6-methylquinoline
4-Hydroxy-6-methylquinoline
Formula: C₁₀H₉NO
MW: 159,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272343
CAS Number: 23432-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR