7-Chloro-4-hydroxy-2-methylquinoline

Supplier: Apollo Scientific

OR307855-1G OR307855-5G
APOSOR307855-1GEA 293 EUR
APOSOR307855-1G APOSOR307855-5G
7-Chloro-4-hydroxy-2-methylquinoline
7-Chloro-4-hydroxy-2-methylquinoline
Formula: C₁₀H₈ClNO
MW: 193,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272320
CAS Number: 15644-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR