6-Bromo-2-chloro-3-(3-chloropropyl)quinoline

Supplier: Apollo Scientific

OR307899-1G OR307899-5G
APOSOR307899-1GEA 310 EUR
APOSOR307899-1G APOSOR307899-5G
6-Bromo-2-chloro-3-(3-chloropropyl)quinoline
6-Bromo-2-chloro-3-(3-chloropropyl)quinoline
Formula: C₁₂H₁₀BrCl₂N
MW: 319,024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09030009
CAS Number: 612494-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR