2-Chloro-6-ethoxyquinoline-3-carbonitrile
Supplier: Apollo Scientific
OR307919-1G
OR307919-5G
APOSOR307919-1GEA
301
EUR
APOSOR307919-1G
APOSOR307919-5G
2-Chloro-6-ethoxyquinoline-3-carbonitrile
2-Chloro-6-ethoxyquinoline-3-carbonitrile
Formula:
C₁₂H₉ClN₂O MW: 232,666 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09035791 CAS Number: 937672-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...