2-Chloro-6-ethylquinoline-3-carbonitrile

Supplier: Apollo Scientific

OR307921-1G
APOSOR307921-1GEA 557 EUR
APOSOR307921-1G
2-Chloro-6-ethylquinoline-3-carbonitrile
2-Chloro-6-ethylquinoline-3-carbonitrile
Formula: C₁₂H₉ClN₂
MW: 216,666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03047545
CAS Number: 498548-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR