1-Methylcyclohexane-1-carboxaldehyde, Technical Grade
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Formyl-1-methylcyclohexane
OR30798-1G
OR30798-5G
OR30798-10G
APOSOR30798-1GEA
157
EUR
APOSOR30798-1G
APOSOR30798-5G
APOSOR30798-10G
1-Methylcyclohexane-1-carboxaldehyde, Technical Grade
1-Methylcyclohexane-1-carboxaldehyde
Formula:
C₈H₁₄O Storage Temperature: Refrigerator |
MDL Number:
MFCD12028616 CAS Number: 6140-64-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...