5-(Hydroxymethyl)-2-methyl-2H-indazole 95%

Supplier: Apollo Scientific
Warning

OR30866-100MG OR30866-250MG
APOSOR30866-100MGEA 50.9 EUR
APOSOR30866-100MG APOSOR30866-250MG
5-(Hydroxymethyl)-2-methyl-2H-indazole 95%
5-(Hydroxymethyl)-2-methyl-2H-indazole
Formula: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD12028587
CAS Number: 1159511-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR