6-Acetyl-1-methyl-1H-indazole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(1-Methyl-1H-indazol-6-yl)ethan-1-one, 1-(1-Methyl-1H-indazol-6-yl)ethanone, 6-Ethanoyl-1-methyl-1H-indazole
OR30890-250MG
OR30890-50MG
APOSOR30890-250MGEA
63.8
EUR
APOSOR30890-250MG
APOSOR30890-50MG
6-Acetyl-1-methyl-1H-indazole
6-Acetyl-1-methyl-1H-indazole
Formula:
C₁₀H₁₀N₂O MW: 174.199 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11869768 CAS Number: 1159511-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...