8-Bromo-4-hydroxy-2-methylquinoline

Supplier: Apollo Scientific

Synonyms: 8-Bromo-2-methylquinolin-4-ol

OR309232-5G
APOSOR309232-5GEA 180 EUR
APOSOR309232-5G
8-Bromo-4-hydroxy-2-methylquinoline
8-Bromo-4-hydroxy-2-methylquinoline
Formula: C₁₀H₈BrNO
MW: 238,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00484434
CAS Number: 1201-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR