8-Chloro-4-hydroxyquinoline-3-carboxylic acid
Supplier: Apollo Scientific
OR309234-1G
OR309234-5G
APOSOR309234-1GEA
293
EUR
APOSOR309234-1G
APOSOR309234-5G
8-Chloro-4-hydroxyquinoline-3-carboxylic acid
8-Chloro-4-hydroxyquinoline-3-carboxylic acid
Formula:
C₁₀H₆ClNO₃ MW: 223,613 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00973835 CAS Number: 35966-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...