8-Ethyl-4-hydroxyquinoline-3-carboxylic acid
Supplier: Apollo Scientific
OR309237-1G
OR309237-5G
APOSOR309237-1GEA
310
EUR
APOSOR309237-1G
APOSOR309237-5G
8-Ethyl-4-hydroxyquinoline-3-carboxylic acid
8-Ethyl-4-hydroxyquinoline-3-carboxylic acid
Formula:
C₁₂H₁₁NO₃ MW: 217,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02323074 CAS Number: 63136-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...