2-Chloro-7-methylquinoline-3-methanol

Supplier: Apollo Scientific

OR309266-1G OR309266-5G
APOSOR309266-1GEA 317 EUR
APOSOR309266-1G APOSOR309266-5G
2-Chloro-7-methylquinoline-3-methanol
2-Chloro-7-methylquinoline-3-methanol
Formula: C₁₁H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD02232326
CAS Number: 170848-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR