2-Chloro-8-ethylquinoline-3-methanol

Supplier: Apollo Scientific

OR309267-1G OR309267-5G
APOSOR309267-1GEA 325 EUR
APOSOR309267-1G APOSOR309267-5G
2-Chloro-8-ethylquinoline-3-methanol
2-Chloro-8-ethylquinoline-3-methanol
Formula: C₁₂H₁₂ClNO
MW: 221,683 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998024
CAS Number: 1017429-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR