6-Bromo-2,8-dimethylquinoline-3-carboxylic acid ethyl ester

Supplier: Apollo Scientific

OR309318-1G OR309318-5G
APOSOR309318-1GEA 335 EUR
APOSOR309318-1G APOSOR309318-5G
6-Bromo-2,8-dimethylquinoline-3-carboxylic acid ethyl ester
6-Bromo-2,8-dimethylquinoline-3-carboxylic acid ethyl ester
Formula: C₁₄H₁₄BrNO₂
MW: 308,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12675068
CAS Number: 1189106-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR