6-Methylquinazoline-2-carboxylic acid hydrochloride
Supplier: Apollo Scientific
OR309395-1G
OR309395-250MG
APOSOR309395-1GEA
799
EUR
APOSOR309395-1G
APOSOR309395-250MG
6-Methylquinazoline-2-carboxylic acid hydrochloride
6-Methylquinazoline-2-carboxylic acid hydrochloride
Formula:
C₁₀H₈N₂O₂·ClH MW: 224,644 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12675147 CAS Number: 1204812-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...