6-Bromo-5-methylpyridine-3-boronic acid pinacol ester
Supplier: Apollo Scientific
OR310013-1G
OR310013-500MG
APOSOR310013-1GEA
248
EUR
APOSOR310013-1G
APOSOR310013-500MG
6-Bromo-5-methylpyridine-3-boronic acid pinacol ester
6-Bromo-5-methylpyridine-3-boronic acid pinacol ester
Formula:
C₁₂H₁₇BBrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08063089 CAS Number: 1256360-64-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...