2-Chloro-4-(4-chlorophenoxy)benzaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR31006-1G
OR31006-250MG
APOSOR31006-1GEA
543
EUR
APOSOR31006-1G
APOSOR31006-250MG
2-Chloro-4-(4-chlorophenoxy)benzaldehyde
2-Chloro-4-(4-chlorophenoxy)benzaldehyde
Formula:
C₁₃H₈Cl₂O₂ MW: 267,107 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11557797 CAS Number: 1092294-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...