N-Benzyl-4-bromoaniline
Supplier: Apollo Scientific
OR310080-500MG
OR310080-100MG
APOSOR310080-500MGEA
319
EUR
APOSOR310080-500MG
APOSOR310080-100MG
N-Benzyl-4-bromoaniline
N-Benzyl-4-bromoaniline
Formula:
C₁₃H₁₂BrN MW: 262,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01427002 CAS Number: 2879-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...