6-Chloro-2-methylnicotinic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Chloro-2-methylpyridine-3-carboxylic acid
OR310139-1G
OR310139-250MG
APOSOR310139-1GEA
168
EUR
APOSOR310139-1G
APOSOR310139-250MG
6-Chloro-2-methylnicotinic acid
6-Chloro-2-methylnicotinic acid
Formula:
C₇H₆ClNO₂ MW: 171,581 g/mol Melting Pt: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD09842452 CAS Number: 137129-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...