1-Methylnaphthalene-2-boronic acid 97%

Supplier: Apollo Scientific

Synonyms: (1-Methylnaphthalen-2-yl)boronic acid

OR310268-1G
APOSOR310268-1GEA 319 EUR
APOSOR310268-1G
1-Methylnaphthalene-2-boronic acid 97%
1-Methylnaphthalene-2-boronic acid
Formula: C₁₁H₁₁BO₂
MW: 186,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566508
CAS Number: 590401-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR