2’-Bromo-4’-iodoacetophenone 95%

Supplier: Apollo Scientific
Warning

OR31027-1G OR31027-250MG OR31027-5G
APOSOR31027-1GEA 633 EUR
APOSOR31027-1G APOSOR31027-250MG APOSOR31027-5G
2’-Bromo-4’-iodoacetophenone 95%
2’-Bromo-4’-iodoacetophenone
Formula: C₈H₆BrIO
MW: 324,941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391606
CAS Number: 1261816-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR