(2R)-2-(4-Bromophenyl)-2-hydroxyacetic acid 99+% e.e.
Supplier: Apollo Scientific
|
Warning
|
OR310337-1G
OR310337-500MG
APOSOR310337-1GEA
1070
EUR
APOSOR310337-1G
APOSOR310337-500MG
(2R)-2-(4-Bromophenyl)-2-hydroxyacetic acid 99+% e.e.
(2R)-2-(4-Bromophenyl)-2-hydroxyacetic acid
Formula:
C₈H₇BrO₃ Melting Pt: 115…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00798431 CAS Number: 32189-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...