2-Methoxy-6-nitrophenol
Supplier: Apollo Scientific
|
Warning
|
OR310383-250MG
OR310383-1G
OR310383-5G
APOSOR310383-250MGEA
69.8
EUR
APOSOR310383-250MG
APOSOR310383-1G
APOSOR310383-5G
2-Methoxy-6-nitrophenol
2-Methoxy-6-nitrophenol
Formula:
C₇H₇NO₄ MW: 169,135 g/mol Melting Pt: 62…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD08693430 CAS Number: 15969-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...