2-Amino-5-bromo-3-methoxybenzoic acid
Supplier: Apollo Scientific
OR310923-1G
APOSOR310923-1GEA
0
EUR
APOSOR310923-1G
2-Amino-5-bromo-3-methoxybenzoic acid
2-Amino-5-bromo-3-methoxybenzoic acid
Formula:
C₈H₈BrNO₃ MW: 246,058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11101005 CAS Number: 864293-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...