[(5-Bromopyridin-2-yl)carbamothioyl]carbamic acid

Supplier: Apollo Scientific

OR310924-1G OR310924-500MG
APOSOR310924-1GEA 873 EUR
APOSOR310924-1G APOSOR310924-500MG
[(5-Bromopyridin-2-yl)carbamothioyl]carbamic acid
[(5-Bromopyridin-2-yl)carbamothioyl]carbamic acid
Formula: C₇H₆BrN₃O₂S
MW: 276,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28101533
CAS Number: 1620482-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR