2-[(2Z)-Azepan-2-ylidene]-2-cyanoacetamide

Supplier: Apollo Scientific

OR310996-1G
APOSOR310996-1GEA 780 EUR
APOSOR310996-1G
2-[(2Z)-Azepan-2-ylidene]-2-cyanoacetamide
2-[(2Z)-Azepan-2-ylidene]-2-cyanoacetamide
Formula: C₉H₁₃N₃O
MW: 179,219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20754284
CAS Number: 76140-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR