3-[4-(Benzyloxy)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole
Supplier: Apollo Scientific
OR311003-1G
APOSOR311003-1GEA
780
EUR
APOSOR311003-1G
3-[4-(Benzyloxy)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole
3-[4-(Benzyloxy)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole
Formula:
C₁₆H₁₃ClN₂O₂ MW: 300,74 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09034182 CAS Number: 937650-48-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...